accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFNOO

CAS RN 446-36-6

IUPAC Name: 5-fluoro-2-nitrophenol
Synonyms: 5-fluoro-2-nitrophenol
Molecular Weight (g/mol): 157.1
Molecular Formula: C6H4FNO3
InChi Key: QQURWFRNETXFTN-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)