Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-nitrophenol, 98%

Catalog No. AAA1256830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1256830 250 g
AAA1256809 10 g
AAA1256818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1256830 Supplier Thermo Scientific Chemicals Supplier No. A1256830
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 446-36-6 | C6H4FNO3 | 157.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-36-6
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00007107
InChI Key QQURWFRNETXFTN-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorophenol, 3-fluoro-6-nitrophenol, phenol, 5-fluoro-2-nitro, 4-fluoro-2-hydroxynitrobenzene, 5-fluoro-2-nitro-phenol, pubchem1490, pubchem2371, acmc-209jzc, 3-fluoro 6-nitrophenol, 5-flouro-2-nitrophenol
PubChem CID 9937
IUPAC Name 5-fluoro-2-nitrophenol
SMILES C1=CC(=C(C=C1F)O)[N+](=O)[O-]

Specifications

Melting Point 30°C to 36°C
Boiling Point 260°C
Flash Point 91°C (195°F)
Quantity 250 g
Beilstein 1870311
Formula Weight 157.1
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 207-168-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.