accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4461-33-0

IUPAC Name: benzoyl isocyanate
Synonyms: benzoyl isocyanate
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: LURYMYITPCOQAU-UHFFFAOYSA-N
SMILES: O=C=NC(=O)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)