Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl isocyanate, tech. 90%, Thermo Scientific™

Catalog No. AAA1158306
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1158306 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1158306 Supplier Thermo Scientific Chemicals Supplier No. A1158306

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4461-33-0
Density 1.171
Boiling Point 48°C to 49°C (0.9mmHg)
Flash Point 72°C (161°F)
Odor Pungent
Molecular Formula C8H5NO2
Refractive Index 1.551
Quantity 5g
MDL Number MFCD00002032
UN Number UN2206
Beilstein 878595
Sensitivity Moisture sensitive
Synonym benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
InChI Key LURYMYITPCOQAU-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)N=C=O
IUPAC Name benzoyl isocyanate
Molecular Weight (g/mol) 147.133
PubChem CID 78210
Formula Weight 147.13
Percent Purity 90%
Grade Technical
Chemical Name or Material Benzoyl isocyanate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 224-717-5
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.