accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4461-41-0

IUPAC Name: (2Z)-2-chlorobut-2-ene
Synonyms: (2Z)-2-chlorobut-2-ene
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: DSDHFHLZEFQSFM-ARJAWSKDSA-N
SMILES: C\C=C(\C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results