Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-butene 96.0+%, TCI America™
SDP

Catalog No. C16225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16225ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16225ML Supplier TCI America Supplier No. C16225ML

Chemical Identifiers

CAS 4461-41-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000860
InChI Key DSDHFHLZEFQSFM-ARJAWSKDSA-N
Synonym 1-Methyl-1-propenyl Chloride
PubChem CID 5463022
IUPAC Name (2Z)-2-chlorobut-2-ene
SMILES C\C=C(\C)Cl

Specifications

Boiling Point 63°C
Quantity 5 mL
UN Number 1993
Formula Weight 90.55
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-butene
EINECSNumber (2)-0117
RTECSNumber EM4265000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.