accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONOO

CAS RN 446838-34-2

IUPAC Name: N-benzyl-4-nitro-N-phenylbenzamide
Synonyms: N-benzyl-4-nitro-N-phenylbenzamide
Molecular Weight (g/mol): 332.36
Molecular Formula: C20H16N2O3
InChi Key: QFYBLCAEBPADFC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results