accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 446866-83-7

IUPAC Name: 3-[(morpholin-4-yl)methyl]benzaldehyde
Synonyms: 3-(morpholin-4-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: KMYNYFKOOXFQGB-UHFFFAOYSA-N
SMILES: O=CC1=CC(CN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results