CAS RN 4472-45-1
IUPAC Name:
4-chloro-2,6-dimethylpyrimidine
Synonyms:
4-chloro-2,6-dimethylpyrimidine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
GSXFOGXQLRLSKK-UHFFFAOYSA-N
SMILES:
CC1=CC(Cl)=NC(C)=N1
1
–
1
of
1
results
1
Promotions Available
4-Chloro-2,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals
4-Chloro-2,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals
CAS: 4472-45-1 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.586 MDL Number: MFCD00234197 InChI Key: GSXFOGXQLRLSKK-UHFFFAOYSA-N PubChem CID: 3154199 IUPAC Name: 4-chloro-2,6-dimethylpyrimidine SMILES: CC1=CC(=NC(=N1)C)Cl