accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4472-92-8

IUPAC Name: 2-[(2E)-3-phenylprop-2-en-1-ylidene]propanedioic acid
Synonyms: 2-[(2E)-3-phenylprop-2-en-1-ylidene]propanedioic acid
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: BLPUXJIIRIWMSQ-QPJJXVBHSA-N
SMILES: OC(=O)C(=C\C=C\C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results