accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SSCH3H3CCH3SCH3H3CCH3SCH3H3CCH3

CAS RN 447463-65-2

IUPAC Name: 1,2,4,5-tetrakis(tert-butylsulfanyl)benzene
Synonyms: 1,2,4,5-tetrakis(tert-butylsulfanyl)benzene
Molecular Weight (g/mol): 430.79
Molecular Formula: C22H38S4
InChi Key: LOCUYYPERSWXJI-UHFFFAOYSA-N
SMILES: CC(C)(C)SC1=CC(SC(C)(C)C)=C(SC(C)(C)C)C=C1SC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results