accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 44767-62-6

IUPAC Name: chloro(hexyl)magnesium
Synonyms: chloro(hexyl)magnesium
Molecular Weight (g/mol): 144.93
Molecular Formula: C6H13ClMg
InChi Key: ODOOOEXDBNGOOF-UHFFFAOYSA-M
SMILES: CCCCCC[Mg]Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results