Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylmagnesium chloride, 2M solution in THF, AcroSeal™

Catalog No. AC434688000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434688000 800 mL AcroSeal™ Glass Bottle
AC434681000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434688000 Supplier Thermo Scientific Chemicals Supplier No. 434688000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 44767-62-6 | C6H13ClMg | 144.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44767-62-6 , 109-99-9
Molecular Formula C6H13ClMg
Molecular Weight (g/mol) 144.93
MDL Number MFCD06798087
InChI Key ODOOOEXDBNGOOF-UHFFFAOYSA-M
Synonym hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10877247
SMILES CCCCCC[Mg]Cl

Specifications

Boiling Point 65.0°C to 68.0°C
Color Brown to Brown-Gray
Physical Form Solution
Quantity 800 mL
Flash Point −25°C
Solubility Information Solubility in water: reacts
IUPAC Name magnesium;hexane;chloride
Formula Weight 144.93
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.965
Density 0.9650g/mL
Chemical Name or Material Hexylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.