accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 448-39-5

IUPAC Name: N-(4-fluoro-2-nitrophenyl)acetamide
Synonyms: N-(4-fluoro-2-nitrophenyl)acetamide
Molecular Weight (g/mol): 198.15
Molecular Formula: C8H7FN2O3
InChi Key: UZBZEUCQENVPQB-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results