Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Fluoro-2'-nitroacetanilide 98.0+%, TCI America™
SDP

Catalog No. F028425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F028425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F028425G Supplier TCI America Supplier No. F028425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 448-39-5
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.153
MDL Number MFCD00031517
InChI Key UZBZEUCQENVPQB-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-nitroacetanilide, n-4-fluoro-2-nitrophenyl acetamide, 4-fluoro-2-nitroacetanilide, maybridge1_003501, 4/'-fluoro-2/'-nitroacetanilide, labotest-bb lt00455473, labotest-bb lt03332301, acetamide,n-4-fluoro-2-nitrophenyl, n-4-fluoro-2-nitro-phenyl-acetamide, n-4-fluoro-2-nitrophenyl acetamide #
PubChem CID 136289
IUPAC Name N-(4-fluoro-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)F)[N+](=O)[O-]

Specifications

Melting Point 72°C
Color Yellow
Quantity 25 g
Formula Weight 198.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Fluoro-2′-nitroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.