accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3

CAS RN 4481-30-5

IUPAC Name: (3-methylbutan-2-yl)benzene
Synonyms: benzene, (1,2-dimethylpropyl)-
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: NQRMTOKLHZNAQH-UHFFFAOYNA-N
SMILES: CC(C)C(C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results