Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,2-Dimethylpropyl)benzene 98.0+%, TCI America™
SDP

Catalog No. D136801ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D136801ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D136801ML Supplier TCI America Supplier No. D13680.1ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4481-30-5
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00059213
InChI Key NQRMTOKLHZNAQH-UHFFFAOYNA-N
Synonym 2-Methyl-3-phenylbutane
PubChem CID 98361
IUPAC Name (3-methylbutan-2-yl)benzene
SMILES CC(C)C(C)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 188°C
Quantity 0.1 mL
UN Number 3295
Formula Weight 148.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (1,2-Dimethylpropyl)benzene
EINECSNumber (3)-0022
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.