accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiClClCl

CAS RN 4484-72-4

IUPAC Name: trichloro(dodecyl)silane
Synonyms: silane, trichlorododecyl-
Molecular Weight (g/mol): 303.77
Molecular Formula: C12H25Cl3Si
InChi Key: BNCXNUWGWUZTCN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)