Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyltrichlorosilane, 96%, Thermo Scientific™

Catalog No. AAA1317836
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1317836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1317836 Supplier Thermo Scientific Chemicals Supplier No. A1317836

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
n-Dodecyltrichlorosilane is employed in the medicinal field, electronic industry and used as polymer material. It is a coupling and reducing agent. It is also used as a lubricant.

Solubility
Soluble in parts of organic solvents.

Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4484-72-4
Molecular Formula C12H25Cl3Si
Molecular Weight (g/mol) 303.767
MDL Number MFCD00000483
InChI Key BNCXNUWGWUZTCN-UHFFFAOYSA-N
Synonym dodecyltrichlorosilane, trichloro dodecyl silane, silane, trichlorododecyl, dodecyl trichlorosilane, silane, dodecyltrichloro, n-dodecyltrichlorosilane, dodecyl trichlorosilane dot, hsdb 387, trichlorolaurylsilane, acmc-1ai7g
PubChem CID 20568
IUPAC Name trichloro(dodecyl)silane
SMILES CCCCCCCCCCCC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 155°C (10mmHg)
Quantity 500 g
Linear Formula CH3(CH2)11SiCl3
UN Number UN1771
Beilstein 1758004
Flash Point 165°C (329°F)
Solubility Information Soluble in parts of organic solvents.
Formula Weight 303.78
Percent Purity 96%
Refractive Index 1.458
Sensitivity Moisture sensitive
Density 1.024
Chemical Name or Material n-Dodecyltrichlorosilane
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DODECYLTRICHLOROSILANE
EINECSNumber 224-769-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.