accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 448912-61-6

IUPAC Name: N-(2-ethylhexyl)-3-nitrobenzamide
Synonyms: N-(2-ethylhexyl)-3-nitrobenzamide
Molecular Weight (g/mol): 278.35
Molecular Formula: C15H22N2O3
InChi Key: CYGCGKPZQVHKCK-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results