accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 4497-05-6

IUPAC Name: 1-phenyltetradecan-1-one
Synonyms: 1-phenyltetradecan-1-one
Molecular Weight (g/mol): 288.48
Molecular Formula: C20H32O
InChi Key: LXUIUVLDNRQBQJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results