Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecanophenone 98.0+%, TCI America™
SDP

Catalog No. T08045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08045G 5 g
T080425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08045G Supplier TCI America Supplier No. T08045G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4497-05-6
Molecular Formula C20H32O
Molecular Weight (g/mol) 288.48
MDL Number MFCD00008985
InChI Key LXUIUVLDNRQBQJ-UHFFFAOYSA-N
Synonym Myristophenone, Phenyl Tridecyl Ketone
PubChem CID 78248
IUPAC Name 1-phenyltetradecan-1-one
SMILES CCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 206°C
Quantity 5 g
Formula Weight 288.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetradecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.