accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 449758-17-2

IUPAC Name: 1-(oxan-2-yl)-1H-pyrazole
Synonyms: 1-(oxan-2-yl)pyrazole
Molecular Weight (g/mol): 152.2
Molecular Formula: C8H12N2O
InChi Key: IMZWSOSYNFVECD-UHFFFAOYNA-N
SMILES: C1CCC(OC1)N1C=CC=N1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)