Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Tetrahydropyranyl)-1H-pyrazole, 98%, Thermo Scientific™

Catalog No. AAH6424603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6424603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6424603 Supplier Thermo Scientific Chemicals Supplier No. H6424603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 449758-17-2
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD11044692
InChI Key IMZWSOSYNFVECD-UHFFFAOYSA-N
Synonym 1-2-tetrahydropyranyl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 1-oxan-2-yl-1h-pyrazole, 1-oxan-2-yl pyrazole, 1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl, 1-2-tetrahydropyranyl pyrazole, 1-tetrahydro-pyran-2-yl-1h-pyrazole, 1-2-oxanyl pyrazole, ksc588k5l, 1-tetrahydropyran-2-ylpyrazole
PubChem CID 21866310
IUPAC Name 1-(oxan-2-yl)pyrazole
SMILES C1CCOC(C1)N2C=CC=N2

Specifications

Color Colorless to Yellow
Density 1.084 g/mL
Boiling Point 60°C to 65°C (0.8 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.503
Quantity 1 g
Beilstein 9694988
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.