accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NHOClNOO

CAS RN 449794-75-6

IUPAC Name: 4-chloro-N-(2-ethylhexyl)-3-nitrobenzamide
Synonyms: 4-chloro-N-(2-ethylhexyl)-3-nitrobenzamide
Molecular Weight (g/mol): 312.79
Molecular Formula: C15H21ClN2O3
InChi Key: XOGLBFUDOWUBEC-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results