accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 4500-58-7

IUPAC Name: (2-ethylphenyl)sulfanide
Synonyms: (2-ethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: ABROBCBIIWHVNS-UHFFFAOYSA-M
SMILES: CCC1=CC=CC=C1[S-]
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)