Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbenzenethiol 95.0+%, TCI America™
SDP

Catalog No. E07025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07025G 5 g
E070225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07025G Supplier TCI America Supplier No. E07025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4500-58-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010022
InChI Key ABROBCBIIWHVNS-UHFFFAOYSA-N
Synonym 2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
PubChem CID 3734338
IUPAC Name 2-ethylbenzenethiol
SMILES CCC1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 205°C
Quantity 5 g
Formula Weight 138.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylbenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.