CAS RN 45019-28-1
IUPAC Name:
(4R)-4-methylnonanoate
Synonyms:
(4R)-4-methylnonanoate
Molecular Weight (g/mol):
171.26
Molecular Formula:
C10H19O2
InChi Key:
WQTZCQIRCYSUBQ-SECBINFHSA-M
SMILES:
CCCCC[C@@H](C)CCC([O-])=O
1
–
1
of
1
results
1
4-Methylnonanoic Acid 97.0+%, TCI America™
CAS: 45019-28-1 Molecular Formula: C10H20O2 Molecular Weight (g/mol): 172.268 MDL Number: MFCD00051789 InChI Key: WQTZCQIRCYSUBQ-UHFFFAOYSA-N PubChem CID: 62003 IUPAC Name: 4-methylnonanoic acid SMILES: CCCCCC(C)CCC(=O)O