Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylnonanoic Acid 97.0+%, TCI America™
SDP

Catalog No. M23805ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23805ML 5 mL
M238025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23805ML Supplier TCI America Supplier No. M23805ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 45019-28-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00051789
InChI Key WQTZCQIRCYSUBQ-UHFFFAOYSA-N
PubChem CID 62003
IUPAC Name 4-methylnonanoic acid
SMILES CCCCCC(C)CCC(=O)O

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 mL
UN Number 3265
Synonym nonanoic acid, 4-methyl, 4-methylpelargonic acid, 4-methyl nonanoic acid, 4-methylnonan-1-oic acid, fema no. 3574, nonanoic acid, 4-methyl-, 4r, methylpelargonic acid, acmc-20mr8v, nonanoic acid,4-methyl, acmc-1aqz1
Formula Weight 172.27
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methylnonanoic Acid
EINECSNumber (2)-0608
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.