accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNOOH

CAS RN 450368-25-9

IUPAC Name: 2-(phenylamino)pyrimidine-5-carboxylic acid
Synonyms: 2-(phenylamino)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 215.21
Molecular Formula: C11H9N3O2
InChi Key: ZPIYMRAJXNMUPE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(NC2=CC=CC=C2)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results