accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3CH2

CAS RN 45103-58-0

IUPAC Name: 2-(2-methoxyethoxy)ethyl 2-methylprop-2-enoate
Synonyms: methoxydiglycol methacrylate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: DAVVKEZTUOGEAK-UHFFFAOYSA-N
SMILES: COCCOCCOC(=O)C(C)=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results