accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 451485-76-0

IUPAC Name: methyl 4-(1,1-dioxo-1λ⁶-thiomorpholin-4-yl)benzoate
Synonyms: methyl 4-(1,1-dioxo-1λ⁶-thiomorpholin-4-yl)benzoate
Molecular Weight (g/mol): 269.32
Molecular Formula: C12H15NO4S
InChi Key: IMLAPXMJROBNSS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)N1CCS(=O)(=O)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results