CAS RN 451491-47-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Histone Acetyltransferase Activator, CTB
The Histone Acetyltransferase Activator, CTB, also referenced under CAS 451491-47-7, modulates the biological activity of Histone. This small molecule/inhibitor is primarily used for Cell Structure applications.