Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Acetyltransferase Activator, CTB

Catalog No. 38212025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-212-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-212-025MG Supplier MilliporeSigma™ Supplier No. 38212025MG
Add to Cart
Add to Cart

The Histone Acetyltransferase Activator, CTB, also referenced under CAS 451491-47-7, modulates the biological activity of Histone. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
CAS 451491-47-7
Molecular Formula C16H13ClF3NO2
Formula Weight 343.7g/mol
Solubility EtOH
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.