accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNOOHOHO

CAS RN 4515-21-3

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Molecular Weight (g/mol): 267.24
Molecular Formula: C12H13NO6
InChi Key: XYXYXSKSTZAEJW-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results