Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-aspartic Acid 95.0+%, TCI America™
SDP

Catalog No. C06311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C06311G Supplier TCI America Supplier No. C06311G

Specifications

CAS 4515-21-3
Melting Point 115°C
Color White-Yellow
Molecular Formula C12H13NO6
MDL Number MFCD00063181
Quantity 1 g
Synonym z-dl-asp-oh, n-carbobenzoxy-dl-aspartic acid, z-dl-aspartic acid, cbz-dl-aspartic acid, 2-benzyloxy carbonyl amino succinic acid, n-cbz-dl-aspartic acid, n-benzyloxy carbonyl aspartic acid, 2-benzyloxycarbonylamino butanedioic acid, 2-phenylmethoxycarbonylamino butanedioic acid, 2-benzyloxy carbonyl amino butanedioic acid
InChI Key XYXYXSKSTZAEJW-UHFFFAOYNA-N
SMILES OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}butanedioic acid
Molecular Weight (g/mol) 267.24
PubChem CID 70855
Formula Weight 267.24
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-aspartic Acid
Show More Show Less
EINECSNumber (9)-1549
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.