accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FF

CAS RN 452-67-5

IUPAC Name: 1,4-difluoro-2-methylbenzene
Synonyms: 1,4-difluoro-2-methylbenzene
Molecular Weight (g/mol): 128.12
Molecular Formula: C7H6F2
InChi Key: YSNVKDGEALPJGC-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results