Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorotoluene 98.0+%, TCI America™
SDP

Catalog No. D404225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D404225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D404225G Supplier TCI America Supplier No. D404225G

missing translation for 'validMainframeMsg'

Specifications

CAS 452-67-5
Melting Point -35°C
Color Yellow
Boiling Point 117°C
Molecular Formula C7H6F2
Quantity 25 g
MDL Number MFCD00000347
UN Number 1993
Synonym 2,5-difluorotoluene, benzene, 1,4-difluoro-2-methyl, 1,4-difluoro-2-methyl-benzene, 1,4-difluor-2-methylbenzol, 2,5-difluorotolune, 2,5-difluoro toluene, p-ditrifluorotoluene, acmc-209k1g, ksc236k9r, benzene,4-difluoro-2-methyl
InChI Key YSNVKDGEALPJGC-UHFFFAOYSA-N
SMILES CC1=CC(F)=CC=C1F
IUPAC Name 1,4-difluoro-2-methylbenzene
Molecular Weight (g/mol) 128.12
PubChem CID 67980
Formula Weight 128.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.