CAS RN 452-81-3
CAS RN 452-81-3
IUPAC Name:
2-fluoro-4-methylphenol
Synonyms:
2-fluoro-4-methylphenol
Molecular Weight (g/mol):
126.13
Molecular Formula:
C7H7FO
InChi Key:
WJKISRFVKIOBCQ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(O)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 2-fluoro-4-methylphenol
Sigma Aldrich 2-fluoro-4-methylphenol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.