accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 45206-91-5

IUPAC Name: hexadecan-7-one
Synonyms: hexadecan-7-one
Molecular Weight (g/mol): 240.43
Molecular Formula: C16H32O
InChi Key: XMIMKDVDMTZIIW-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)CCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)