Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hexadecanone, 98+%, Thermo Scientific™

Catalog No. AAB2510322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2510322 100 g
AAB2510306 5 g
AAB2510314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2510322 Supplier Thermo Scientific Chemicals Supplier No. B2510322

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45206-91-5
Molecular Formula C16H32O
Molecular Weight (g/mol) 240.431
MDL Number MFCD00027354
InChI Key XMIMKDVDMTZIIW-UHFFFAOYSA-N
Synonym 7-hexadecanone, acmc-1as9q
PubChem CID 98679
IUPAC Name hexadecan-7-one
SMILES CCCCCCCCCC(=O)CCCCCC

Specifications

Melting Point 36°C to 38°C
Quantity 100 g
Formula Weight 240.43
Percent Purity ≥98%
Chemical Name or Material 7-Hexadecanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.