accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4525-44-4

H3CSiCH3ClCl

CAS RN 4525-44-4

IUPAC Name: dichloro(ethyl)methylsilane
Synonyms: dichloro-ethyl-methyl silane
Molecular Weight (g/mol): 143.08
Molecular Formula: C3H8Cl2Si
InChi Key: PNECSTWRDNQOLT-UHFFFAOYSA-N
SMILES: CC[Si](C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results