Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmethyldichlorosilane, 97%, Thermo Scientific™

Catalog No. AC432890250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432890250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432890250 Supplier Thermo Scientific Chemicals Supplier No. 432890250

Chemical Identifiers

CAS 4525-44-4
Molecular Formula C3H8Cl2Si
Molecular Weight (g/mol) 143.08
MDL Number MFCD00013607
InChI Key PNECSTWRDNQOLT-UHFFFAOYSA-N
Synonym ethylmethyldichlorosilane, methylethyldichlorosilane, dichloro ethyl methylsilane, silane, dichloroethylmethyl, dichloro ethyl methyl silane, unii-47rob570jw, acmc-20akvy, dichloro-ethyl-methyl-silane, dichloro ethyl ?methylsilane, dichloro-ethyl-methyl silane
PubChem CID 78286
IUPAC Name dichloro-ethyl-methylsilane
SMILES CC[Si](C)(Cl)Cl

Specifications

Color Colorless
Density 1.0630g/mL
Boiling Point 100.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 1.063
Formula Weight 143.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethylmethyldichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call
EINECSNumber 224-860-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.