accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4525-75-1

CH3OOCl

CAS RN 4525-75-1

IUPAC Name: 2-chlorophenyl acetate
Synonyms: 2-chlorophenyl acetate
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: CJPVPOYTTALCNX-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results