Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. C212025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C212025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C212025G Supplier TCI America Supplier No. C212025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4525-75-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD01632281
InChI Key CJPVPOYTTALCNX-UHFFFAOYSA-N
Synonym o-acetoxychlorobenzene, o-chlorophenyl acetate, acetic acid, 2-chlorophenyl ester, acetic acid, o-chlorophenyl ester, phenol, o-chloro-, acetate, o-chlorfenylester kyseliny octove, acetic acid 2-chlorophenyl ester, o-chlorfenylester kyseliny octove czech, chlorophenol acetate
PubChem CID 20620
IUPAC Name (2-chlorophenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1Cl

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 103°C
Quantity 25 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorophenyl Acetate
RTECSNumber AG0700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.