accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4526-93-6

IUPAC Name: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanamido)acetate
Synonyms: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanamido)acetate
Molecular Weight (g/mol): 324.33
Molecular Formula: C15H20N2O6
InChi Key: ZOONJGSLKPDXCV-UHFFFAOYNA-N
SMILES: CCOC(=O)CNC(=O)C(CO)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results