accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHNBr

CAS RN 452972-09-7

IUPAC Name: (5-bromopyridin-3-yl)boronic acid
Synonyms: 5-bromopyridin-3-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: ICCGFOKNFZWCTJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results