Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals

Catalog No. AAL2008403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2008403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2008403 Supplier Thermo Scientific Chemicals Supplier No. L2008403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 452972-09-7 | Molecular Formula: C5H5BBrNO2 | Molecular Weight: 201.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452972-09-7
Molecular Formula C5H5BBrNO2
Molecular Weight (g/mol) 201.81
MDL Number MFCD02685634
InChI Key ICCGFOKNFZWCTJ-UHFFFAOYSA-N
Synonym 5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305
PubChem CID 3814811
IUPAC Name (5-bromopyridin-3-yl)boronic acid
SMILES OB(O)C1=CN=CC(Br)=C1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 9119873
Formula Weight 201.82
Percent Purity 85%
Grade Technical
Chemical Name or Material 5-Bromopyridine-3-boronic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.