accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOF

CAS RN 453-18-9

IUPAC Name: methyl 2-fluoroacetate
Synonyms: fluoressigsaeure-methylester
Molecular Weight (g/mol): 92.07
Molecular Formula: C3H5FO2
InChi Key: RJBYSQHLLIHSLT-UHFFFAOYSA-N
SMILES: COC(=O)CF

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results