Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Fluoroacetate 99.0+%, TCI America™
SDP

Catalog No. F018325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F018325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F018325G Supplier TCI America Supplier No. F018325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 453-18-9
Melting Point -40°C
Color Yellow
Boiling Point 104°C
Molecular Formula C3H5FO2
Quantity 25 g
MDL Number MFCD00040096
UN Number 2929
Synonym Fluoroacetic Acid Methyl Ester
InChI Key RJBYSQHLLIHSLT-UHFFFAOYSA-N
SMILES COC(=O)CF
IUPAC Name methyl 2-fluoroacetate
Molecular Weight (g/mol) 92.069
PubChem CID 9959
Formula Weight 92.07
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Fluoroacetate
Show More Show Less
RTECSNumber AH8400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.